Assay Details
| Gene Symbol: CDA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127515.4 | NP_179547.1 | 1 | 739 | 89 | |
| GenBank mRNA | AF134487.1 | - | 1 | 739 | 89 | |
| AJ005261.1 | - | 1 | 664 | 89 | ||
| AY128366.1 | - | 1 | 677 | 89 | ||
| BT008708.1 | - | 1 | 598 | 89 | ||
More Information
Set Membership: |
|

CDA1
Genomic Map