Assay Details
| Gene Symbol: PRMT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127525.5 | NP_179557.1 | 4 - 5 | 580 | 113 | |
| GenBank mRNA | AK227449.1 | - | 4 - 5 | 436 | 113 | |
| BT006491.1 | - | 4 - 5 | 400 | 113 | ||
More Information
Set Membership: |
|

PRMT1A
Genomic Map