Assay Details
| Gene Symbol: MIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127538.4 | NP_565459.1 | 3 - 4 | 685 | 131 | |
| GenBank mRNA | BT033135.1 | - | 3 - 4 | 260 | 131 | |
More Information
Set Membership: |
|

MIOX2
Genomic Map