Assay Details
| Gene Symbol: NADP-ME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335646.1 | NP_001324890.1 | 13 - 14 | 1149 | 64 | |
| NM_127548.4 | NP_179580.1 | 15 - 16 | 1522 | 64 | ||
| GenBank mRNA | AY062734.1 | - | 15 - 16 | 1416 | 64 | |
| BT003371.1 | - | 15 - 16 | 1366 | 64 | ||
More Information
Set Membership: |
|

NADP-ME1
Genomic Map