Assay Details
| Gene Symbol: NADP-ME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127548.4 | NP_179580.1 | 2 - 3 | 401 | 103 | |
| GenBank mRNA | AY062734.1 | - | 2 - 3 | 295 | 103 | |
| BT003371.1 | - | 2 - 3 | 245 | 103 | ||
More Information
Set Membership: |
|

NADP-ME1
Genomic Map