Assay Details
| Gene Symbol: RIN13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127590.6 | NP_179621.1 | 2 - 3 | 702 | 69 | |
| GenBank mRNA | BT033116.1 | - | 2 - 3 | 413 | 69 | |
More Information
Set Membership: |
|
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIN13| Gene Symbol: RIN13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127590.6 | NP_179621.1 | 2 - 3 | 702 | 69 | |
| GenBank mRNA | BT033116.1 | - | 2 - 3 | 413 | 69 | |
Set Membership: |
|
Genomic Map