Assay Details
| Gene Symbol: AT2G20420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127601.4 | NP_179632.1 | 7 - 8 | 922 | 132 | |
| GenBank mRNA | AJ001808.1 | - | 7 - 8 | 774 | 132 | |
| AY099707.1 | - | 7 - 8 | 774 | 132 | ||
| AY128889.1 | - | 7 - 8 | 690 | 132 | ||
More Information
Set Membership: |
|

AT2G20420
Genomic Map