Assay Details
| Gene Symbol: BME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115338.1 | NP_191041.1 | 2 - 3 | 508 | 91 | |
| NM_180373.3 | NP_850704.1 | 2 - 3 | 529 | 91 | ||
| GenBank mRNA | AF412107.1 | - | 2 - 3 | 508 | 91 | |
| AK317503.1 | - | 2 - 3 | 429 | 91 | ||
| AY078029.1 | - | 1 - 2 | 231 | 91 | ||
More Information
Set Membership: |
|

BME3
Genomic Map