Assay Details
| Gene Symbol: UBA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339827.1 | NP_001325973.1 | 1 - 2 | 1602 | 97 | |
| NM_001339828.1 | NP_001319772.1 | 1 - 2 | 1602 | 97 | ||
| NM_001339829.1 | NP_001325971.1 | 1 - 2 | 1636 | 97 | ||
| NM_180379.4 | NP_850710.1 | 1 - 2 | 1636 | 97 | ||
| GenBank mRNA | AJ439404.1 | - | 1 - 2 | 1459 | 96 | |
| AY037245.1 | - | 1 - 2 | 1539 | 97 | ||
More Information
Set Membership: |
|

UBA2A
Genomic Map