Assay Details
| Gene Symbol: MTHFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115860.4 | NP_191556.1 | 6 - 7 | 1247 | 65 | |
| NM_180392.1 | NP_850723.1 | 6 - 7 | 1190 | 65 | ||
| NM_180393.1 | NP_850724.1 | 6 - 7 | 1190 | 65 | ||
| GenBank mRNA | AF181966.1 | - | 6 - 7 | 1170 | 65 | |
| AY070034.1 | - | 6 - 7 | 1190 | 65 | ||
| AY122922.1 | - | 6 - 7 | 1108 | 65 | ||
More Information
Set Membership: |
|

MTHFR1
Genomic Map