Assay Details
| Gene Symbol: MAP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035821.1 | NP_001030898.1 | 11 - 12 | 1285 | 135 | |
| NM_001203210.1 | NP_001190139.1 | 11 - 12 | 1521 | 135 | ||
| NM_115862.3 | NP_567089.1 | 11 - 12 | 1312 | 135 | ||
| NM_180394.2 | NP_850725.1 | 10 - 11 | 1452 | 135 | ||
| GenBank mRNA | AF300880.1 | - | 10 - 11 | 1377 | 135 | |
| AY084710.1 | - | 11 - 12 | 1311 | 135 | ||
More Information
Set Membership: |
|

MAP2B
Genomic Map