Assay Details
| Gene Symbol: MAP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035821.1 | NP_001030898.1 | 2 - 3 | 236 | 110 | |
| NM_001203210.1 | NP_001190139.1 | 2 - 3 | 472 | 110 | ||
| NM_115862.3 | NP_567089.1 | 2 - 3 | 263 | 110 | ||
| NM_180394.2 | NP_850725.1 | 1 - 2 | 403 | 110 | ||
| GenBank mRNA | AF300880.1 | - | 1 - 2 | 328 | 110 | |
| AY084710.1 | - | 2 - 3 | 262 | 110 | ||
More Information
Set Membership: |
|

MAP2B
Genomic Map