Assay Details
| Gene Symbol: MAP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035821.1 | NP_001030898.1 | 5 - 6 | 494 | 60 | |
| NM_001203210.1 | NP_001190139.1 | 5 - 6 | 730 | 60 | ||
| NM_115862.3 | NP_567089.1 | 5 - 6 | 521 | 60 | ||
| NM_180394.2 | NP_850725.1 | 4 - 5 | 661 | 60 | ||
| GenBank mRNA | AF300880.1 | - | 4 - 5 | 586 | 60 | |
| AY084710.1 | - | 5 - 6 | 520 | 60 | ||
More Information
Set Membership: |
|

MAP2B
Genomic Map