Assay Details
| Gene Symbol: MAP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035821.1 | NP_001030898.1 | 6 - 7 | 636 | 132 | |
| NM_001203210.1 | NP_001190139.1 | 6 - 7 | 872 | 132 | ||
| NM_115862.3 | NP_567089.1 | 6 - 7 | 663 | 132 | ||
| NM_180394.2 | NP_850725.1 | 5 - 6 | 803 | 132 | ||
| GenBank mRNA | AF300880.1 | - | 5 - 6 | 728 | 132 | |
| AY084710.1 | - | 6 - 7 | 662 | 132 | ||
More Information
Set Membership: |
|

MAP2B
Genomic Map