Assay Details
Gene Symbol: GLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120496.3 | NP_568134.1 | 15 - 16 | 2757 | 111 | |
NM_180432.2 | NP_850763.1 | 14 - 15 | 2589 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
