Assay Details
Gene Symbol: GLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120496.3 | NP_568134.1 | 29 - 30 | 4555 | 97 | |
NM_180432.2 | NP_850763.1 | 28 - 29 | 4387 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
