Assay Details
| Gene Symbol: SSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085103.1 | NP_001078572.1 | 3 - 4 | 496 | 66 | |
| NM_001343218.1 | NP_001318546.1 | 3 - 4 | 761 | 66 | ||
| NM_121224.4 | NP_196747.1 | 2 - 3 | 1079 | 66 | ||
| NM_180478.1 | NP_850809.1 | 3 - 4 | 635 | 66 | ||
| NM_203038.3 | NP_974767.1 | 3 - 4 | 574 | 66 | ||
| GenBank mRNA | AY085441.1 | - | 2 - 3 | 530 | 66 | |
| BT020406.1 | - | 1 - 2 | 354 | 66 | ||
More Information
Set Membership: |
|

SSP5
Genomic Map