Assay Details
| Gene Symbol: CHR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121867.4 | NP_568365.2 | 21 - 22 | 2812 | 84 | |
| NM_180516.2 | NP_850847.1 | 21 - 22 | 2810 | 84 | ||
More Information
Set Membership: |
|

CHR17
Genomic Map