Assay Details
| Gene Symbol: CHR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121867.4 | NP_568365.2 | 3 - 4 | 563 | 67 | |
| NM_180516.2 | NP_850847.1 | 3 - 4 | 561 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHR17| Gene Symbol: CHR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121867.4 | NP_568365.2 | 3 - 4 | 563 | 67 | |
| NM_180516.2 | NP_850847.1 | 3 - 4 | 561 | 67 | ||
Set Membership: |
|