Assay Details
| Gene Symbol: CHR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121867.4 | NP_568365.2 | 6 - 7 | 947 | 77 | |
| NM_180516.2 | NP_850847.1 | 6 - 7 | 945 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHR17| Gene Symbol: CHR17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121867.4 | NP_568365.2 | 6 - 7 | 947 | 77 | |
| NM_180516.2 | NP_850847.1 | 6 - 7 | 945 | 77 | ||
Set Membership: |
|