Assay Details
| Gene Symbol: PYR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036865.1 | NP_001031942.1 | 6 - 7 | 668 | 118 | |
| NM_001343973.1 | NP_001332394.1 | 6 - 7 | 885 | 118 | ||
| NM_180536.3 | NP_850867.1 | 6 - 7 | 709 | 118 | ||
| NM_180537.3 | NP_850868.1 | 6 - 7 | 722 | 118 | ||
| GenBank mRNA | AF000147.1 | - | 6 - 7 | 590 | 118 | |
| BT029445.1 | - | 5 - 6 | 530 | 118 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Detect Genomic DNA  Arabidopsis thaliana  Probe spans exons | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 PYR6
PYR6 Genomic Map
Genomic Map Back To Top
 Back To Top