Assay Details
| Gene Symbol: AGL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180557.2 | NP_850888.2 | 1 | 636 | 88 | |
| GenBank mRNA | AY141248.1 | - | 1 | 636 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL34| Gene Symbol: AGL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180557.2 | NP_850888.2 | 1 | 636 | 88 | |
| GenBank mRNA | AY141248.1 | - | 1 | 636 | 88 | |
Set Membership: |
|