Assay Details
| Gene Symbol: RPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180572.2 | NP_850903.1 | 1 - 2 | 379 | 90 | |
| GenBank mRNA | AF370232.1 | - | 1 - 2 | 341 | 90 | |
| AY059914.1 | - | 1 - 2 | 361 | 90 | ||
| AY088230.1 | - | 1 - 2 | 380 | 90 | ||
| AY142540.1 | - | 1 - 2 | 304 | 90 | ||
More Information
Set Membership: |
|

RPS1
Genomic Map