Assay Details
| Gene Symbol: CER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197961.1 | NP_001184890.1 | 5 - 6 | 1186 | 96 | |
| NM_001331327.1 | NP_001320548.1 | 4 - 5 | 1317 | 96 | ||
| NM_001331328.1 | NP_001320547.1 | 3 - 4 | 1931 | 96 | ||
| NM_100101.4 | NP_171723.2 | 5 - 6 | 1518 | 96 | ||
| NM_180601.2 | NP_850932.2 | 4 - 5 | 1312 | 96 | ||
| GenBank mRNA | AK226895.1 | - | 4 - 5 | 1296 | 96 | |
More Information
Set Membership: |
|

CER1
Genomic Map