Assay Details
| Gene Symbol: SR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331358.1 | NP_001321307.1 | 12 - 13 | 1570 | 76 | |
| NM_001331359.1 | NP_001321308.1 | 12 - 13 | 1508 | 76 | ||
| NM_180602.2 | NP_850933.1 | 12 - 13 | 1103 | 76 | ||
| NM_180603.1 | NP_850934.1 | 12 - 13 | 1315 | 76 | ||
| GenBank mRNA | AK118379.1 | - | 12 - 13 | 1000 | 76 | |
| AY085920.1 | - | 12 - 13 | 1001 | 76 | ||
| AY128338.1 | - | 12 - 13 | 1309 | 76 | ||
More Information
Set Membership: |
|

SR34
Genomic Map