Assay Details
| Gene Symbol: SR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331357.1 | NP_001318908.1 | 5 - 6 | 510 | 143 | |
| NM_001331358.1 | NP_001321307.1 | 5 - 6 | 512 | 143 | ||
| NM_001331359.1 | NP_001321308.1 | 5 - 6 | 450 | 143 | ||
| NM_180602.2 | NP_850933.1 | 5 - 6 | 556 | 143 | ||
| NM_180603.1 | NP_850934.1 | 5 - 6 | 415 | 143 | ||
| GenBank mRNA | AK118379.1 | - | 5 - 6 | 453 | 143 | |
| AY085920.1 | - | 5 - 6 | 454 | 143 | ||
| AY128338.1 | - | 5 - 6 | 409 | 143 | ||
| BT006316.1 | - | 4 - 5 | 361 | 143 | ||
More Information
Set Membership: |
|

SR34
Genomic Map