Assay Details
Gene Symbol: OBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035911.2 | NP_001030988.1 | 2 | 1634 | 136 | |
NM_100637.4 | NP_563792.3 | 2 | 1398 | 136 | ||
NM_180607.2 | NP_850938.1 | 1 | 1494 | 136 | ||
GenBank mRNA | AY062715.1 | - | 1 | 1259 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
