Assay Details
| Gene Symbol: SCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198248.1 | NP_001185177.1 | 16 - 17 | 1969 | 71 | |
| NM_001333364.1 | NP_001320175.1 | 16 - 17 | 1984 | 71 | ||
| NM_103797.4 | NP_175333.3 | 16 - 17 | 1921 | 71 | ||
| NM_180628.4 | NP_850959.1 | 16 - 17 | 2007 | 71 | ||
| GenBank mRNA | AY082605.1 | - | 16 - 17 | 1814 | 71 | |
More Information
Set Membership: |
|

SCD1
Genomic Map