Assay Details
| Gene Symbol: ARFA1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334452.1 | NP_001323062.1 | 4 - 5 | 908 | 64 | |
| NM_001334453.1 | NP_001323063.1 | 4 - 5 | 897 | 64 | ||
| NM_001334454.1 | NP_001323060.1 | 4 - 5 | 881 | 64 | ||
| NM_001334455.1 | NP_001323061.1 | 4 - 5 | 784 | 64 | ||
| NM_105717.3 | NP_177206.1 | 4 - 5 | 449 | 64 | ||
| NM_180644.3 | NP_850975.1 | 4 - 5 | 475 | 64 | ||
| NM_202391.2 | NP_974120.1 | 5 - 6 | 553 | 64 | ||
| Gene Symbol: ARF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF360164.1 | - | 4 - 5 | 423 | 64 | |
| AY056341.1 | - | 3 - 4 | 340 | 64 | ||
| AY087235.1 | - | 4 - 5 | 472 | 64 | ||
More Information
Set Membership: |
|

ARFA1D
Genomic Map