Assay Details
| Gene Symbol: ARFA1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334452.1 | NP_001323062.1 | 6 - 7 | 1103 | 82 | |
| NM_001334453.1 | NP_001323063.1 | 6 - 7 | 1092 | 82 | ||
| NM_001334454.1 | NP_001323060.1 | 6 - 7 | 1076 | 82 | ||
| NM_001334455.1 | NP_001323061.1 | 6 - 7 | 979 | 82 | ||
| NM_105717.3 | NP_177206.1 | 6 - 7 | 644 | 82 | ||
| NM_180644.3 | NP_850975.1 | 6 - 7 | 670 | 82 | ||
| NM_202391.2 | NP_974120.1 | 7 - 8 | 748 | 82 | ||
| Gene Symbol: ARF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF360164.1 | - | 6 - 7 | 618 | 82 | |
| AY056341.1 | - | 5 - 6 | 535 | 82 | ||
| AY087235.1 | - | 6 - 7 | 667 | 82 | ||
More Information
Set Membership: |
|

ARFA1D
Genomic Map