Assay Details
| Gene Symbol: ELF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120506.5 | NP_196044.2 | 6 - 7 | 2419 | 76 | |
| GenBank mRNA | AY664500.1 | - | 6 - 7 | 2028 | 76 | |
More Information
Set Membership: |
|

ELF6
Genomic Map