Assay Details
| Gene Symbol: ZAT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120516.4 | NP_196054.1 | 1 | 728 | 60 | |
| GenBank mRNA | AF022658.1 | - | 1 | 445 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZAT6| Gene Symbol: ZAT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120516.4 | NP_196054.1 | 1 | 728 | 60 | |
| GenBank mRNA | AF022658.1 | - | 1 | 445 | 60 | |
Set Membership: |
|