Assay Details
Gene Symbol: NAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120523.4 | NP_196061.1 | 2 - 3 | 738 | 84 | |
GenBank mRNA | AY099609.1 | - | 2 - 3 | 611 | 84 | |
AY128859.1 | - | 2 - 3 | 442 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
