Assay Details
Gene Symbol: NAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120523.4 | NP_196061.1 | 4 - 5 | 1494 | 108 | |
GenBank mRNA | AY099609.1 | - | 4 - 5 | 1367 | 108 | |
AY128859.1 | - | 4 - 5 | 1198 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
