Assay Details
| Gene Symbol: UBC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085061.1 | NP_001078530.1 | 3 - 4 | 408 | 105 | |
| NM_001342801.1 | NP_001332674.1 | 3 - 4 | 715 | 105 | ||
| NM_001342802.1 | NP_001332673.1 | 3 - 4 | 621 | 105 | ||
| NM_120590.3 | NP_568148.1 | 3 - 4 | 444 | 105 | ||
| GenBank mRNA | AF428320.1 | - | 3 - 4 | 416 | 105 | |
| AY088106.1 | - | 3 - 4 | 440 | 105 | ||
| AY116948.1 | - | 1 - 2 | 145 | 105 | ||
| DQ027036.1 | - | 1 - 2 | 145 | 105 | ||
More Information
Set Membership: |
|

UBC22
Genomic Map