Assay Details
| Gene Symbol: UBC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085061.1 | NP_001078530.1 | 4 - 5 | 490 | 162 | |
| NM_001342801.1 | NP_001332674.1 | 4 - 5 | 797 | 162 | ||
| NM_001342802.1 | NP_001332673.1 | 4 - 5 | 703 | 162 | ||
| NM_120590.3 | NP_568148.1 | 4 - 5 | 526 | 162 | ||
| GenBank mRNA | AF428320.1 | - | 4 - 5 | 498 | 162 | |
| AY088106.1 | - | 4 - 5 | 522 | 162 | ||
| AY116948.1 | - | 2 - 3 | 227 | 162 | ||
| DQ027036.1 | - | 2 - 3 | 227 | 162 | ||
More Information
Set Membership: |
|

UBC22
Genomic Map