Assay Details
| Gene Symbol: RPN8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120660.4 | NP_196197.1 | 4 - 5 | 586 | 114 | |
| GenBank mRNA | AF378878.1 | - | 4 - 5 | 582 | 114 | |
| AY230840.1 | - | 4 - 5 | 454 | 114 | ||
| BT002610.1 | - | 4 - 5 | 454 | 114 | ||
More Information
Set Membership: |
|

RPN8A
Genomic Map