Assay Details
| Gene Symbol: RPN8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203304.1 | NP_001190233.1 | 7 - 8 | 845 | 76 | |
| NM_120660.4 | NP_196197.1 | 7 - 8 | 849 | 76 | ||
| GenBank mRNA | AF378878.1 | - | 7 - 8 | 845 | 76 | |
| AY087393.1 | - | 7 - 8 | 846 | 76 | ||
| AY230839.1 | - | 7 - 8 | 777 | 76 | ||
| AY230840.1 | - | 7 - 8 | 717 | 76 | ||
| BT002610.1 | - | 7 - 8 | 717 | 76 | ||
| U54560.2 | - | 7 - 8 | 791 | 76 | ||
More Information
Set Membership: |
|

RPN8A
Genomic Map