Assay Details
| Gene Symbol: RPN8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203304.1 | NP_001190233.1 | 9 - 10 | 987 | 75 | |
| NM_120660.4 | NP_196197.1 | 9 - 10 | 991 | 75 | ||
| GenBank mRNA | AF378878.1 | - | 9 - 10 | 987 | 75 | |
| AY087393.1 | - | 9 - 10 | 988 | 75 | ||
| AY230839.1 | - | 9 - 10 | 919 | 75 | ||
| AY230840.1 | - | 9 - 10 | 859 | 75 | ||
| BT002610.1 | - | 9 - 10 | 859 | 75 | ||
| U54560.2 | - | 9 - 10 | 933 | 75 | ||
More Information
Set Membership: |
|

RPN8A
Genomic Map