Assay Details
Gene Symbol: ROPGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342847.1 | NP_001332731.1 | 2 - 3 | 605 | 75 | |
NM_120676.3 | NP_196213.2 | 2 - 3 | 605 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
