Assay Details
| Gene Symbol: 2-Cys Prx B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342875.1 | NP_001331225.1 | 1 - 2 | 435 | 70 | |
| NM_120712.4 | NP_568166.1 | 1 - 2 | 534 | 70 | ||
| GenBank mRNA | AF326871.1 | - | 1 - 2 | 241 | 70 | |
| AF339693.1 | - | 1 - 2 | 234 | 70 | ||
| AY054621.1 | - | 1 - 2 | 366 | 70 | ||
| AY081503.1 | - | 1 - 2 | 234 | 70 | ||
More Information
Set Membership: |
|

2-Cys Prx B
Genomic Map