Assay Details
| Gene Symbol: RBL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125710.1 | NP_001119182.1 | 4 - 5 | 837 | 87 | |
| NM_120807.4 | NP_196342.1 | 4 - 5 | 1027 | 87 | ||
| GenBank mRNA | AY058157.1 | - | 4 - 5 | 978 | 87 | |
| AY098983.1 | - | 4 - 5 | 923 | 87 | ||
More Information
Set Membership: |
|

RBL3
Genomic Map