Assay Details
| Gene Symbol: GRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342946.1 | NP_001318499.1 | 1 - 2 | 315 | 142 | |
| NM_120833.4 | NP_196368.2 | 1 - 2 | 320 | 142 | ||
| GenBank mRNA | DQ446928.1 | - | 1 - 2 | 254 | 142 | |
More Information
Set Membership: |
|

GRP14
Genomic Map