Assay Details
| Gene Symbol: GRP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125713.1 | NP_001119185.1 | 1 - 2 | 450 | 71 | |
| NM_001342948.1 | NP_001331377.1 | 1 - 2 | 454 | 71 | ||
| NM_120835.3 | NP_196370.1 | 1 - 2 | 450 | 71 | ||
| GenBank mRNA | AK229852.1 | - | 1 - 2 | 450 | 71 | |
More Information
Set Membership: |
|

GRP17
Genomic Map