Assay Details
| Gene Symbol: SMC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120848.2 | NP_196383.1 | 15 - 16 | 1898 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMC6A| Gene Symbol: SMC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120848.2 | NP_196383.1 | 15 - 16 | 1898 | 86 | |
Set Membership: |
|