Assay Details
| Gene Symbol: SNAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342964.1 | NP_001318503.1 | 2 - 3 | 500 | 63 | |
| NM_120870.2 | NP_196405.1 | 1 - 2 | 408 | 63 | ||
| GenBank mRNA | DQ056673.1 | - | 1 - 2 | 345 | 63 | |
More Information
Set Membership: |
|

SNAP29
Genomic Map