Assay Details
| Gene Symbol: HEMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120911.4 | NP_196445.1 | 1 - 2 | 480 | 65 | |
| GenBank mRNA | AF419614.1 | - | 1 - 2 | 380 | 65 | |
| AY070431.1 | - | 1 - 2 | 349 | 65 | ||
| AY123037.1 | - | 1 - 2 | 250 | 65 | ||
More Information
Set Membership: |
|

HEMC
Genomic Map