Assay Details
Gene Symbol: SULA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335740.1 | NP_001324767.1 | 1 - 2 | 182 | 65 | |
NM_127700.4 | NP_565505.1 | 1 - 2 | 113 | 65 | ||
GenBank mRNA | AF324701.2 | - | 1 - 2 | 100 | 65 | |
AF326895.1 | - | 1 - 2 | 100 | 65 | ||
AF339713.1 | - | 1 - 2 | 85 | 65 | ||
AJ516948.1 | - | 1 - 2 | 85 | 65 | ||
BT000757.1 | - | 1 - 2 | 100 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
