Assay Details
| Gene Symbol: XK-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335744.1 | NP_001323495.1 | 4 - 5 | 735 | 88 | |
| NM_001335745.1 | NP_001323494.1 | 4 - 5 | 748 | 88 | ||
| NM_127708.3 | NP_179732.2 | 4 - 5 | 748 | 88 | ||
| NM_201776.3 | NP_973505.1 | 4 - 5 | 735 | 88 | ||
| GenBank mRNA | AK175657.1 | - | 4 - 5 | 669 | 88 | |
| AK176783.1 | - | 4 - 5 | 669 | 88 | ||
| AY136397.1 | - | 4 - 5 | 671 | 88 | ||
| BT000210.1 | - | 4 - 5 | 656 | 88 | ||
More Information
Set Membership: |
|

XK-1
Genomic Map