Assay Details
| Gene Symbol: VHA-A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127713.4 | NP_179736.1 | 14 - 15 | 2112 | 67 | |
| GenBank mRNA | AF424556.1 | - | 14 - 15 | 1961 | 67 | |
| AY099690.1 | - | 14 - 15 | 2004 | 67 | ||
More Information
Set Membership: |
|

VHA-A2
Genomic Map